MilliporeSigma
  • Comparative study of X-ray treatment and photodynamic therapy by using 5-aminolevulinic acid conjugated gold nanoparticles in a melanoma cell line
Comparative study of X-ray treatment and photodynamic therapy by using 5-aminolevulinic acid conjugated gold nanoparticles in a melanoma cell line
Artificial Cells, Nanomedicine, and Biotechnology (Print) (2017)
Mohammadi Z, et al