MilliporeSigma
  • Synthesis, Structures and Antimicrobial Activities of Two Cobalt(II) Complexes [Co(L1)2(OH2)2] and [Co(L2)2].
Synthesis, Structures and Antimicrobial Activities of Two Cobalt(II) Complexes [Co(L1)2(OH2)2] and [Co(L2)2].
Acta chimica Slovenica (2017-04-06)
Yong-Jun Han, Li Wang, Qing-Bin Li, Ling-Wei Xue
ABSTRACT
A new cobalt(II) complex, [Co(L1)2(OH2)2] (1), was prepared by the reaction of 3-bromo-5-chlorosalicylaldehyde (HL1) with cobalt nitrate in methanol. Reaction of 1 with cyclopropylamine in methanol afforded the Schiff base cobalt(II) complex, [Co(L2)2] (2), where L2 is the deprotonated form of 2-bromo-4-chloro-6-(cyclopropyliminomethyl)phenol (HL2). The complexes have been characterized by elemental analyses, IR spectroscopy, and single-crystal X-ray diffraction. The L1 ligand coordinates to the Co atom through the phenolate O and carbonyl O atoms, while the L2 ligand coordinates to the Co atom through the phenolate O and imino N atoms. The Co atom in complex 1 adopts octahedral coordination and that in complex 2 adopts tetrahedral coordination. The effect of the free ligands and the cobalt complexes on the antimicrobial activities against Staphylococcus aureus, Escherichia coli, and Candida albicans was studied.
Sigma-Aldrich
Cyclopropylamine, 98%