MilliporeSigma
  • Allium cepa L. Response to Sodium Selenite (Se(IV)) Studied in Plant Roots by a LC-MS-Based Proteomic Approach.
Allium cepa L. Response to Sodium Selenite (Se(IV)) Studied in Plant Roots by a LC-MS-Based Proteomic Approach.
Journal of agricultural and food chemistry (2017-05-04)
Jakub Karasinski, Kazimierz Wrobel, Alma Rosa Corrales Escobosa, Anna Konopka, Ewa Bulska, Katarzyna Wrobel
ABSTRACT
Liquid chromatography-high-resolution mass spectrometry was used for the first time to investigate the impact of Se(IV) (10 mgSe L
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
5-Methylcytidine, ≥99%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM