MilliporeSigma
  • Effect of paroxetine on left ventricular remodeling in an in vivo rat model of myocardial infarction.
Effect of paroxetine on left ventricular remodeling in an in vivo rat model of myocardial infarction.
Basic research in cardiology (2017-03-30)
Thomas Ravn Lassen, Jan Møller Nielsen, Jacob Johnsen, Steffen Ringgaard, Hans Erik Bøtker, Steen Buus Kristiansen
ABSTRACT
Left ventricular (LV) remodeling following a myocardial infarction (MI) involves formation of reactive oxygen species (ROS). Paroxetine, a selective serotonin reuptake inhibitor, has an antioxidant effect in the vascular wall. We investigated whether paroxetine reduces myocardial ROS formation and LV remodeling following a MI. In a total of 32 Wistar rats, MI was induced by a 30-min ligation of the left anterior descending artery followed by 7- or 28-day reperfusion. During the 28 days of reperfusion, LV remodeling was evaluated by magnetic resonance imaging (MRI) and echocardiography (n = 20). After 28 days of reperfusion, the susceptibility to ventricular tachycardia was evaluated prior to sacrifice and histological assessment of myocyte cross-sectional area, fibrosis, and presence of myofibroblasts. Myocardial ROS formation was measured with dihydroethidium after 7 days of reperfusion in separate groups (n = 12). Diastolic LV volume, evaluated by MRI (417 ± 60 vs. 511 ± 64 µL, p < 0.05), and echocardiography (515 ± 80 vs. 596 ± 83 µL, p < 0.05) as well as diastolic LV internal diameter evaluated with echocardiography (7.2 ± 0.6 vs. 8.1 ± 0.7 mm, p < 0.05) were lower in the paroxetine group than in controls. Furthermore, myocyte cross-sectional area was reduced in the paroxetine group compared with controls (277 ± 26 vs. 354 ± 23 mm
Product Description
Millipore
2,3,5-Triphenyl-tetrazolium chloride solution, suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC