MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • CD47 promotes ovarian cancer progression by inhibiting macrophage phagocytosis.
CD47 promotes ovarian cancer progression by inhibiting macrophage phagocytosis.
Oncotarget (2017-04-06)
Ran Liu, Huiting Wei, Peng Gao, Hu Yu, Ke Wang, Zheng Fu, Baohui Ju, Meng Zhao, Shangwen Dong, Zhijun Li, Yifeng He, Yuting Huang, Zhi Yao
ABSTRACT
Targeting CD47 efficiently enhances macrophage phagocytosis in both physiological and pathological conditions. Anti-CD47 antibodies have been shown to inhibit the progression of several types of cancer. However, the mechanism of anti-CD47 monoclonal antibody (mAb) treatment remains controversial. In this study, we confirmed that CD47 protein is highly expressed in ovarian cancer, and is correlated with poor clinical characteristics and prognosis. CD47 knockdown in the ovarian cancer cell line, SK-OV-3, promoted phagocytosis by macrophages in vitro and inhibited tumor growth in vivo. These data combined suggest that CD47 inhibition is a potential strategy for cancer treatment. Using an anti-CD47 mAb, we found that CD47 inhibition in both SK-OV-3 cells and primary cancer cells was able to recapitulate our knockdown results and led to an increase in the number of infiltrating macrophages. In addition, the CD133+ tumor initiating cells expressed a high level of CD47, and anti-CD47 mAb treatment was able to trigger the phagocytosis of this cell population. In conclusion, our results indicate that CD47 inhibits macrophage phagocytosis of ovarian cancer cells, and down-regulation of CD47 or inhibiting CD47 by mAb was able to reverse the negative effect. Thus, CD47 antibody therapy may be a promising strategy to treat ovarian cancer.
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
SAFC
Insulin, Human Recombinant, for research or for further manufacturing use, dry powder
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture