MilliporeSigma
Conduction abnormalities during dipyridamole stress testing.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology (2015-10-24)
Samia Massalha, Ilya Reizberg, Ora Israel, Michael Kapeliovich, Haitham Sholy, Amjad Koskosi, Zohar Keidar, Ibrahim Marai
ABSTRACT
Pharmacological stress tests using dipyridamole are considered to be safe. However, cases of atrioventricular (AV) block have been reported. We retrospectively analyzed ECG at baseline and during dipyridamole stress tests of 2010 consecutive patients (patients with second or third degree AV block were excluded). At baseline, 350 (17.4%) patients had conduction abnormalities. Following dipyridamole infusion 16 patients (0.8%) developed a transient change in AV conduction (15 patients) and or sinus arrest (1 patient). Compared to patients without baseline conduction abnormalities, patients with any conduction abnormalities at baseline were at a higher risk for the development of AV block after dipyridamole infusion [0.3% vs 3.14%, respectively; P < .0001].