MilliporeSigma
  • Distinctive role of efferocytosis in dendritic cell maturation and migration in sterile or infectious conditions.
Distinctive role of efferocytosis in dendritic cell maturation and migration in sterile or infectious conditions.
Immunology (2017-03-08)
Letícia de Aquino Penteado, Naiara Naiana Dejani, Felipe Fortino Verdan, Allan Botinhon Orlando, Victoria Eugenia Niño, Fernanda De Nuzzi Dias, Ana Carolina Guerta Salina, Alexandra Ivo Medeiros
ABSTRACT
Efferocytosis, or clearance of apoptotic cells (ACs), by dendritic cells (DCs) leads to immune response suppression and tolerance to self-antigens. However, efferocytosis of infected apoptotic cells (IACs) leads to the production of a mixed pro- and anti-inflammatory cytokine milieu. We examined the DC phenotype and ability to migrate after phagocytosis of ACs or IACs and observed higher levels of CD86 and CCR7 expression in DCs, as well as enhanced migration capacity following efferocytosis of IACs. Interestingly, higher levels of interleukin-1β, interleukin-10 and prostaglandin E
Sigma-Aldrich
Human PLG (Plasminogen) ELISA Kit, for cell culture supernatants, plasma, and serum samples