MilliporeSigma
  • Cost-effectiveness of percutaneous coronary intervention with cobalt-chromium everolimus eluting stents versus bare metal stents: Results from a patient level meta-analysis of randomized trials.
Cost-effectiveness of percutaneous coronary intervention with cobalt-chromium everolimus eluting stents versus bare metal stents: Results from a patient level meta-analysis of randomized trials.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2016-08-17)
Nicole Ferko, Giuseppe Ferrante, James T Hasegawa, Tanya Schikorr, Ireena M Soleas, John B Hernandez, Manel Sabaté, Christoph Kaiser, Salvatore Brugaletta, Jose Maria de la Torre Hernandez, Soeren Galatius, Angel Cequier, Franz Eberli, Adam de Belder, Patrick W Serruys, Marco Valgimigli
ABSTRACT
Second-generation drug eluting stents (DES) may reduce costs and improve clinical outcomes compared to first-generation DES with improved cost-effectiveness when compared to bare metal stents (BMS). We aimed to conduct an economic evaluation of a cobalt-chromium everolimus eluting stent (Co-Cr EES) compared with BMS in percutaneous coronary intervention (PCI). To conduct a cost-effectiveness analysis (CEA) of a cobalt-chromium everolimus eluting stent (Co-Cr EES) versus BMS in PCI. A Markov state transition model with a 2-year time horizon was applied from a US Medicare setting with patients undergoing PCI with Co-Cr EES or BMS. Baseline characteristics, treatment effects, and safety measures were taken from a patient level meta-analysis of 5 RCTs (n = 4,896). The base-case analysis evaluated stent-related outcomes; a secondary analysis considered the broader set of outcomes reported in the meta-analysis. The base-case and secondary analyses reported an additional 0.018 and 0.013 quality-adjusted life years (QALYs) and cost savings of $236 and $288, respectively with Co-Cr EES versus BMS. Results were robust to sensitivity analyses and were most sensitive to the price of clopidogrel. In the probabilistic sensitivity analysis, Co-Cr EES was associated with a greater than 99% chance of being cost saving or cost effective (at a cost per QALY threshold of $50,000) versus BMS. Using data from a recent patient level meta-analysis and contemporary cost data, this analysis found that PCI with Co-Cr EES is more effective and less costly than PCI with BMS. © 2016 The Authors. Catheterization and Cardiovascular Interventions Published by Wiley Periodicals, Inc.
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid