MilliporeSigma
Triterpenoid saponins from the roots of Spergularia marginata.
Phytochemistry (2017-04-23)
David Pertuit, Mustafa Larshini, Malika Aitsidi Brahim, Mohamed Markouk, Anne-Claire Mitaine-Offer, Thomas Paululat, Stéphanie Delemasure, Patrick Dutartre, Marie-Aleth Lacaille-Dubois
ABSTRACT
Phytochemical investigations of the roots of Spergularia marginata had led to the isolation of four previously undescribed triterpenoid saponins, a known one and one spinasterol glycoside. Their structures were established by extensive NMR and mass spectroscopic techniques as 3-O-β-D-glucuronopyranosyl echinocystic acid 28-O-α-L-arabinopyranosyl-(1 → 2)-α-L-rhamnopyranosyl-(1 → 3)-β-D-xylopyranosyl-(1 → 4)-α-L-rhamnopyranosyl-(1 → 2)-α-L- arabinopyranosyl ester, 3-O-β-D-glucopyranosyl-(1 → 3)-β-D-glucuronopyranosyl echinocystic acid 28-O-α-L-arabinopyranosyl-(1 → 2)-α-L-rhamnopyranosyl-(1 → 3)-β-D-xylopyranosyl-(1 → 4)-α-L-rhamnopyranosyl-(1 → 2)- α-L-arabinopyranosyl ester, 3-O-β-D-glucopyranosyl-(1 → 4)-3-O-sulfate-β-D-glucuronopyranosyl echinocystic acid 28-O-α-L-arabinopyranosyl-(1 → 2)-α-L-rhamnopyranosyl-(1 → 3)-β-D-xylopyranosyl-(1 → 4)-α-L-rhamnopyranosyl-(1 → 2)-α-L-arabinopyranosyl ester, and 3-O-β-D-glucopyranosyl-(1 → 4)-β-D-glucuronopyranosyl 21-O-acetyl acacic acid. Their cytotoxicity was evaluated against two human cancer cell lines SW480 and MCF-7. The most active compound showed a cytotoxicity with IC
Sigma-Aldrich
Thiazolidine, 95%