MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Seedless synthesis of nanocomposites, optical properties, and effects of additives on their surface resonance plasmon bands.
Seedless synthesis of nanocomposites, optical properties, and effects of additives on their surface resonance plasmon bands.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-04-14)
Zoya Zaheer, Elham Shafik Aazam
ABSTRACT
The work describes an easy seedless competitive chemical reduction method for the synthesis of Ag@Au/Ag bimetallic nanoparticles by mixing AgNO
Sigma-Aldrich
DL-Cysteine, technical grade