MilliporeSigma
  • Development and validation of a novel multiplexed DNA analysis system, InnoTyper
Development and validation of a novel multiplexed DNA analysis system, InnoTyper
Forensic science international. Genetics (2017-04-10)
Hiromi Brown, Robyn Thompson, Gina Murphy, Dixie Peters, Bobby La Rue, Jonathan King, Anne H Montgomery, Marion Carroll, James Baus, Sid Sinha, Frank R Wendt, Bing Song, Ranajit Chakraborty, Bruce Budowle, Sudhir K Sinha
ABSTRACT
We report here a novel multiplexed DNA analysis system consisting of 20 Alu markers and Amelogenin for analysis of highly degraded forensic biological samples. The key to the success of the system in obtaining results from degraded samples is the primer design yielding small amplicon size (60-125bp) for all 20 markers. The markers included in the InnoTyper
Sigma-Aldrich
Hematin porcine