MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
BID links ferroptosis to mitochondrial cell death pathways.
Redox biology (2017-04-07)
Sandra Neitemeier, Anja Jelinek, Vincenzo Laino, Lena Hoffmann, Ina Eisenbach, Roman Eying, Goutham K Ganjam, Amalia M Dolga, Sina Oppermann, Carsten Culmsee
ABSTRACT
Ferroptosis has been defined as an oxidative and iron-dependent pathway of regulated cell death that is distinct from caspase-dependent apoptosis and established pathways of death receptor-mediated regulated necrosis. While emerging evidence linked features of ferroptosis induced e.g. by erastin-mediated inhibition of the X
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.