MilliporeSigma
  • Determination of flubendiamide in honey at trace levels by using solid phase extraction and liquid chromatography coupled to quadrupole time-of-flight mass spectrometry.
Determination of flubendiamide in honey at trace levels by using solid phase extraction and liquid chromatography coupled to quadrupole time-of-flight mass spectrometry.
Food chemistry (2017-05-12)
Ana M Ares, Silvia Valverde, José L Bernal, Laura Toribio, María J Nozal, José Bernal
ABSTRACT
In this study, a new method has been developed to determine flubendiamide in honey using liquid chromatography coupled to a selective mass spectrometry detector (quadrupole-time-of-flight). An efficient sample treatment involving a solid phase extraction with a C
Product Description
Supelco
Flubendiamide, PESTANAL®, analytical standard
Pricing and availability is not currently available.