MilliporeSigma
  • Probing the Conformation-Dependent Preferential Binding of Ethanol to Cationic Glycylalanylglycine in Water/Ethanol by Vibrational and NMR Spectroscopy.
Probing the Conformation-Dependent Preferential Binding of Ethanol to Cationic Glycylalanylglycine in Water/Ethanol by Vibrational and NMR Spectroscopy.
The journal of physical chemistry. B (2017-05-23)
David DiGuiseppi, Bridget Milorey, Gabrielle Lewis, Nina Kubatova, Stefanie Farrell, Harald Schwalbe, Reinhard Schweitzer-Stenner
ABSTRACT
The conformational propensity of amino acid residues is determined by an intricate balance of peptide-solvent and solvent-solvent interactions. To explore how the systematic replacement of water by a cosolvent affects the solvation of both the amino acid backbone and side chains, we performed a combined vibrational spectroscopy and NMR study of cationic glycylalanylglycine (GAG) in different ethanol/water mixtures of between 0 and 42 mol percent ethanol. Classical model peptide N'-methylacetamide was used as a reference system to probe solvent-induced spectroscopic changes. The alanine residue of GAG in water is known to exhibit a very high propensity for polyproline II (pPII). Adding up to 30 mol % ethanol at room temperature leads only to minor changes in the Ramachandran distribution of alanine, which mostly changes within the individual conformational subspaces. A further increase in the ethanol fractions leads to a destabilization of pPII and a stabilization of β-strand conformations. At higher temperatures, different degrees of enthalpy-entropy compensations lead to a much stronger influence of ethanol on the peptide's conformational distribution. Ethanol-induced changes in chemical shifts and amide I wavenumbers strongly suggest that ethanol replaces water preferentially in the solvation shell of the polar C-terminal peptide group and of the alanine side chain, whereas the N-terminal group remains mostly hydrated. Furthermore, we found that ethanol interacts more strongly with the peptide if the latter adopts β-strand conformations. This leads to an unusual positive temperature coefficient for the chemical shift of the C-terminal amide proton. Our data suggests a picture in which GAG eventually accumulates at water-ethanol interfaces if the ethanol fractions exceed 0.3, which explains why the further addition of ethanol eventually causes self-aggregation and the subsequent formation of a hydrogel.