MilliporeSigma
  • Streptavidin-biotin-peroxidase nanocomplex-amplified microfluidics immunoassays for simultaneous detection of inflammatory biomarkers.
Streptavidin-biotin-peroxidase nanocomplex-amplified microfluidics immunoassays for simultaneous detection of inflammatory biomarkers.
Analytica chimica acta (2017-07-25)
Jing Wu, Yiping Chen, Mingzhu Yang, Yu Wang, Cheng Zhang, Mo Yang, Jiashu Sun, Mengxia Xie, Xingyu Jiang
ABSTRACT
Simultaneous, sensitive and quantitative detection of biomarkers in infectious disease is crucial for guiding antimicrobial treatment and predicting prognosis. This work reported an ultrasensitive and quantitative microfluidic immunoassay combined with the streptavidin-biotin-peroxidase (SA-B-HRP) nanocomplex-signal amplification system (MIS) to detect two inflammatory biomarkers, procalcitonin (PCT, for discriminating bacterial infections from nonbacterial infections) and interleukin-6 (IL-6, for monitoring the kinetics of infectious disease) simultaneously. The amplification system was based on the one step self-assembly of SA and B-HRP to form the SA-B-HRP nanocomplex, which effectively amplified the chemiluminescent signals. The linear ranges for PCT and IL-6 detections by MIS were 250-1.28 × 10
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)