MilliporeSigma
  • Effects of glucose and oxygen deprivation on phosphoinositide hydrolysis in cerebral cortex slices from neonatal rats.
Effects of glucose and oxygen deprivation on phosphoinositide hydrolysis in cerebral cortex slices from neonatal rats.
Life sciences (1996-01-01)
R M Cristòfol, E Rodríguez-Farré, C Sanfeliu
ABSTRACT
The effects of glucose deprivation, hypoxia and glucose-free hypoxia conditions on phosphoinositide (PI) hydrolysis were studied in cortical slices from 8-day-old rats. Only glucose-free hypoxia induced a significant increase of inositol phosphate formation. The inositol phosphate formation induced by noradrenaline, carbachol and several excitatory amino acid receptor agonists, but not the Ca2+ ionophore A23187-induced stimulation, was blocked by glucose-free hypoxia and differentially reduced by glucose and oxygen deprivation depending on the neurotransmitter receptor agonist. The stimulatory effect of glucose-free hypoxia was not reduced by the muscarinic receptor antagonist atropine or by the inhibitors of the excitatory amino acid-stimulated PI hydrolysis DL-2-amino-3-phosphono-propionic acid and L-aspartate-beta-hydroxamate, and neither by the voltage-sensitive Na+ channel tetrodotoxin. The effect of glucose-free hypoxia was partially dependent on extracellular Ca2+ and it was blocked by verapamil and amiloride, but not by nifedipine, Co2+ and neomycin. These results suggest that Ca2+ influx through the Na(+)-Ca2+ exchanger underlies the PI hydrolysis stimulation induced by combined glucose and oxygen deprivation in neonatal cerebral cortical slices.
Sigma-Aldrich
DL-2-Amino-3-phosphonopropionic acid