MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Retrosynthesis of CaCO
Journal of structural biology (2017-05-17)
Andrónico Neira-Carrillo, María Soledad Fernández, Gonzalo Poblete Hevia, José Luis Arias, Denis Gebauer, Helmut Cölfen
ABSTRACT
Gastroliths are highly calcified structures formed in the cardiac stomach wall of crustaceans for the temporary storage of amorphous CaCO
Sigma-Aldrich
Sodium acrylate, 97%