MilliporeSigma
  • A Tumor-pH-Responsive Supramolecular Photosensitizer for Activatable Photodynamic Therapy with Minimal
A Tumor-pH-Responsive Supramolecular Photosensitizer for Activatable Photodynamic Therapy with Minimal
Theranostics (2017-08-19)
Xingshu Li, Bi-Yuan Zheng, Mei-Rong Ke, Yongfan Zhang, Jian-Dong Huang, Juyoung Yoon
ABSTRACT
A major challenge in photodynamic therapy (PDT) is the development of new tumor-targeting photosensitizers. The tumor-specific activation is considered to be an effective strategy for designing these photosensitizers. Herein, we describe a novel tumor-pH-responsive supramolecular photosensitizer, LDH-ZnPcS
Sigma-Aldrich
1,3-Diphenylisobenzofuran, 97%