MilliporeSigma
  • Simultaneous determination of dechloranes, polybrominated diphenyl ethers and novel brominated flame retardants in food and serum.
Simultaneous determination of dechloranes, polybrominated diphenyl ethers and novel brominated flame retardants in food and serum.
Analytical and bioanalytical chemistry (2017-06-09)
Carlos Sales, Giulia Poma, Govindan Malarvannan, Tania Portolés, Joaquin Beltrán, Adrian Covaci
ABSTRACT
A sensitive method for the simultaneous quantification of dechloranes, polybrominated diphenyl ethers (PBDEs) and novel brominated flame retardants (NBFRs) has been developed for gas chromatography (GC) coupled to tandem mass spectrometry operating in electron capture negative ionization (ECNI) mode. The major advance has been achieved by combining selected ion monitoring (SIM) and multiple reaction monitoring (MRM) modes in well-defined time windows, to determine dechloranes, PBDEs and NBFRs at picogram per gram level in one single analysis in complex matrix biological samples. From the chromatographic point of view, efforts were devoted to study several injection modes using multimode inlet (MMI) in order to obtain low instrumental detection limits, necessary for trace compounds such as Dechlorane Plus (DP) isomers. Method performance was also evaluated: calibration curves were linear from 20 fg μL
Sigma-Aldrich
2-Methylheptane, 98%
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)