MilliporeSigma
  • Identification of Keratin 19-Positive Cancer Stem Cells Associating Human Hepatocellular Carcinoma Using
Identification of Keratin 19-Positive Cancer Stem Cells Associating Human Hepatocellular Carcinoma Using
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-09-25)
Takayuki Kawai, Kentaro Yasuchika, Satoru Seo, Tatsuya Higashi, Takamichi Ishii, Yuya Miyauchi, Hidenobu Kojima, Ryoya Yamaoka, Hokahiro Katayama, Elena Yukie Yoshitoshi, Satoshi Ogiso, Sadahiko Kita, Katsutaro Yasuda, Ken Fukumitsu, Yuji Nakamoto, Etsuro Hatano, Shinji Uemoto
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder