MilliporeSigma
  • Modeling of Polarization Transfer Kinetics in Protein Hydration Using Hyperpolarized Water.
Modeling of Polarization Transfer Kinetics in Protein Hydration Using Hyperpolarized Water.
The journal of physical chemistry. B (2017-06-15)
Jihyun Kim, Mengxiao Liu, Christian Hilty
ABSTRACT
Water-protein interactions play a central role in protein structure, dynamics, and function. These interactions, traditionally, have been studied using nuclear magnetic resonance (NMR) by measuring chemical exchange and nuclear Overhauser effect (NOE). Polarization transferred from hyperpolarized water can result in substantial transient signal enhancements of protein resonances due to these processes. Here, we use dissolution dynamic nuclear polarization and flow-NMR for measuring the pH dependence of transferred signals to the protein trypsin. A maximum enhancement of 20 is visible in the amide proton region of the spectrum at pH 6.0, and of 47 at pH 7.5. The aliphatic region is enhanced up to 2.3 times at pH 6.0 and up to 2.5 times at pH 7.5. The time dependence of these observed signals can be modeled quantitatively using rate equations incorporating chemical exchange to amide sites and, optionally, intramolecular NOE to aliphatic protons. On the basis of these two- and three-site models, average exchange (k
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%