MilliporeSigma
Crown Ether Complexes of Alkali-Metal Chlorides from SO
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-05-04)
Kirsten Reuter, Stefan S Rudel, Magnus R Buchner, Florian Kraus, Carsten von Hänisch
ABSTRACT
The structures of alkali-metal chloride SO
Sigma-Aldrich
12-Crown-4, 98%
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis