MilliporeSigma
  • The efficacy and safety of vertebral fracture prevention therapies in post-menopausal osteoporosis treatment: Which therapies work best? a network meta-analysis.
The efficacy and safety of vertebral fracture prevention therapies in post-menopausal osteoporosis treatment: Which therapies work best? a network meta-analysis.
Bone & joint research (2017-08-03)
G Wang, L Sui, P Gai, G Li, X Qi, X Jiang
ABSTRACT
Osteoporosis has become an increasing concern for older people as it may potentially lead to osteoporotic fractures. This study is designed to assess the efficacy and safety of ten therapies for post-menopausal women using network meta-analysis. We conducted a systematic search in several databases, including PubMed and Embase. A random-effects model was employed and results were assessed by the odds ratio (OR) and corresponding 95% confidence intervals (CI). Furthermore, with respect to each outcome, each intervention was ranked according to the surface under the cumulative ranking curve (SUCRA) value. With respect to preventing new vertebral fractures (NVF), all ten drugs outperformed placebo, and etidronate proved to be the most effective treatment (OR 0.24, 95% CI 0.14 to 0.39). In addition, zoledronic acid and parathyroid hormone ranked higher compared with the other drugs. With respect to preventing clinical vertebral fractures (CVF), zoledronic acid proved to be the most effective drug (OR = 0.25, 95% CI 0.08 to 0.92), with denosumab as a desirable second option (OR = 0.48, 95% CI 0.22 to 0.96), when both were compared with placebo. As for adverse events (AE) and severe adverse events (SAE), no significant difference was observed. According to SUCRA, etidronate ranked first in preventing CVF; parathyroid hormone and zoledronic acid ranked highly in preventing NVF and CVF. Raloxifene was safe with a high rank in preventing AEs and SAEs though performed unsatisfactorily in efficacy. This study suggests that, taking efficacy and safety into account, parathyroid hormone and zoledronic acid had the highest probability of satisfactory performance in preventing osteoporotic fractures.