MilliporeSigma
  • Polycyclic aromatic hydrocarbons in urban soil of Novi Sad, Serbia: occurrence and cancer risk assessment.
Polycyclic aromatic hydrocarbons in urban soil of Novi Sad, Serbia: occurrence and cancer risk assessment.
Environmental science and pollution research international (2017-05-26)
Biljana D Škrbić, Nataša Đurišić-Mladenović, Đorđe J Tadić, Jelena Đ Cvejanov
ABSTRACT
Contents of 16 polycyclic aromatic hydrocarbons were analyzed in 30 soil samples from 15 locations in Novi Sad, Serbia, assessing for the first time the corresponding health risks in the Serbian urban zone. Total concentrations were in the range of 22-2247 μg kg