MilliporeSigma
  • Procyanidin B2 gallates inhibit IFN-? and IL-17 production in T cells by suppressing T-bet and ROR?t expression
Procyanidin B2 gallates inhibit IFN-? and IL-17 production in T cells by suppressing T-bet and ROR?t expression
International Immunopharmacology (2017)
Tanaka S, et al.