MilliporeSigma
  • A study of the ?? ? transformation in pure iron: rate variations revealed by means of thermal analysis
A study of the ?? ? transformation in pure iron: rate variations revealed by means of thermal analysis
Philosophical Magazine (2007)
Papandrea C, et al.