MilliporeSigma
  • Evaluation of cell viability dyes in antiviral assays with RNA viruses that exhibit different cytopathogenic properties.
Evaluation of cell viability dyes in antiviral assays with RNA viruses that exhibit different cytopathogenic properties.
Journal of virological methods (2017-04-01)
Donald F Smee, Brett L Hurst, W Joseph Evans, Nathan Clyde, Sean Wright, Christopher Peterson, Kie-Hoon Jung, Craig W Day
ABSTRACT
Studies were conducted to determine the performance of four dyes in assessing antiviral activities of compounds against three RNA viruses with differing cytopathogenic properties. Dyes included alamarBlue
Sigma-Aldrich
6-Azauridine
Pricing and availability is not currently available.