MilliporeSigma
  • Effects of feeding three types of corn-milling coproducts on milk production and ruminal fermentation of lactating Holstein cattle.
Effects of feeding three types of corn-milling coproducts on milk production and ruminal fermentation of lactating Holstein cattle.
Journal of dairy science (2009-09-19)
J M Kelzer, P J Kononoff, A M Gehman, L O Tedeschi, K Karges, M L Gibson
ABSTRACT
Two experiments were conducted to determine the effects of feeding 3 corn-milling coproducts on intake, milk production, ruminal fermentation, and digestibility of lactating Holstein cows. In experiment 1, three corn-milling coproducts were fed at 15% of the diet dry matter (DM) to 28 Holstein cows averaging (+/-SD) 625 +/- 81 kg of body weight and 116 +/- 33 d in milk to determine effects on DM intake and milk production. In experiment 2, the same rations were fed to 4 ruminally fistulated, multiparous Holstein cows averaging 677 +/- 41 kg of body weight and 144 +/- 5 d in milk to determine the effects on ruminal fermentation and digestibility. In both experiments, cows and treatments were assigned randomly in 4 x 4 Latin squares over four 21-d periods. Treatments were formulated by replacing portions of forage and concentrate feeds with 15% coproduct and included 1) 0% coproduct (control), 2) dried distillers grains plus solubles (DDGS), 3) dehydrated corn germ meal (germ), and 4) high-protein dried distillers grains (HPDDG). Feed intake was recorded daily, and milk samples were collected on d 19 to 21 of each period for analysis of major components. Rumen fluid was collected at 10 time points over 24 h post feeding on d 21 of experiment 2. In experiment 1, DM intake was greater for the germ (24.3 kg/d) and DDGS treatments (23.8 kg/d), but DDGS was not different from the control (22.9 kg/d) and HPDDG treatments (22.4 kg/d). Milk production paralleled DM intake and tended to be greater for the germ (32.1 kg/d) and DDGS treatments (30.9 kg/d), but the DDGS treatment was not different from the control (30.6 kg/d) and HPDDG treatments (30.3 kg/d). However, yields of milk fat, milk protein, and 3.5% FCM were similar and averaged (+/-SEM) 1.1 +/- 0.1, 0.9 +/- 0.03, and 31.7 +/- 1.3 kg/d. Milk urea nitrogen was greater for the HPDDG (15.9 mg/dL) and germ treatments (15.5 mg/dL) than for the control (15.0 mg/dL) and DDGS treatments (14.9 mg/dL). In experiment 2, DM intake and milk production were not different across treatments and averaged 26.1 +/- 2.3 and 28.3 +/- 3.9 kg/d. Ruminal pH (6.26 +/- 0.08) and total concentration of volatile fatty acids (125.3 +/- 4.2 mM) were similar. Acetate concentration was higher for the control treatment than the DDGS, germ, and HPDDG treatments (81.7 vs. 75.8, 75.0, and 78.4 mM). Concentrations of propionate and butyrate were not different and averaged 27.8 +/- 1.2 and 14.3 +/- 0.9 mM across treatments. The acetate:propionate ratios for the control, germ, and HPDDG treatments were greater than for the DDGS treatment (3.02, 2.88, and 2.91 vs. 2.62). Dry matter, organic matter, and neutral detergent fiber digestibilities were similar across treatments and averaged 63.5 +/- 2.7, 67.3 +/- 2.2, and 43.5 +/- 4.2%. Milk production followed DM intake in experiment 1, and yield of major milk components was not affected. Results of these experiments indicate that dairy rations can be successfully formulated to include 15% of diet DM as corn-milling coproducts while maintaining or increasing DM intakes and yields of milk and milk components.
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A