MilliporeSigma
  • Monitoring a simple hydrolysis process in an organic solid by observing methyl group rotation.
Monitoring a simple hydrolysis process in an organic solid by observing methyl group rotation.
Solid state nuclear magnetic resonance (2017-03-07)
Peter A Beckmann, Joseph M Bohen, Jamie Ford, William P Malachowski, Clelia W Mallory, Frank B Mallory, Andrew R McGhie, Arnold L Rheingold, Gilbert J Sloan, Steven T Szewczyk, Xianlong Wang, Kraig A Wheeler
ABSTRACT
We report a variety of experiments and calculations and their interpretations regarding methyl group (CH
Sigma-Aldrich
3-Methylglutaric acid, 99%