MilliporeSigma
Spermidine-mediated poly(lactic-
International journal of nanomedicine (2017-09-22)
Jing Tang, Jianming Li, Guo Li, Haitao Zhang, Ling Wang, Dai Li, Jinsong Ding
ABSTRACT
Idiopathic pulmonary fibrosis is a progressive, fatal lung disease with poor survival. The advances made in deciphering this disease have led to the approval of different antifibrotic molecules, such as pirfenidone and nintedanib. An increasing number of studies with particles (liposomes, nanoparticles [NPs], microspheres, nanopolymersomes, and nanoliposomes) modified with different functional groups have demonstrated improvement in lung-targeted drug delivery. In the present study, we prepared, characterized, and evaluated spermidine (Spd)-modified poly(lactic-
Sigma-Aldrich
2-Iminothiolane hydrochloride, ≥98% (TLC), powder