MilliporeSigma
  • Protective Roles of Monsonia angustifolia and Its Active Compounds in Experimental Models of Alzheimer's Disease.
Protective Roles of Monsonia angustifolia and Its Active Compounds in Experimental Models of Alzheimer's Disease.
Journal of agricultural and food chemistry (2017-04-06)
Yoon Sun Chun, Joonki Kim, Sungkwon Chung, Eric Khorombi, Dashnie Naidoo, Rudzani Nthambeleni, Nial Harding, Vinesh Maharaj, Gerda Fouche, Hyun Ok Yang
ABSTRACT
Alzheimer's disease (AD), a progressive neurodegenerative disorder, is characterized by the accumulation of neurotoxic β-amyloid (Aβ) peptides, which consequently affects cognitive decline and memory impairment. Current research on AD treatment is actively focusing on the prevention of neurotoxic Aβ peptide accumulation. Monsonia angustifolia is reported to be consumed as an indigenous vegetable in Tanzania. In this study, we investigated the effect of the ethanol (EtOH) extract of M. angustifolia dried ground material on Aβ production and spatial learning ability as protection against AD. The formation of Aβ peptides was significantly reduced in HeLa cells stably transfected with the Swedish mutant form of β-amyloid precursor protein (APPsw) after treatment with a 60% EtOH extract of M. angustifolia. We next examined the cognitive-improving effects of the EtOH extract in vivo. Tg2576 mice were treated with extract for 6 months and subjected to Morris water maze and novel object recognition tests. The results showed that the 60% EtOH extract of M. angustifolia significantly ameliorated behavioral deficits of the AD transgenic mice and reduced the level of insoluble Aβ42 in the cerebral cortex and hippocampus. We further found that the 60% EtOH extract was effective for memory function recovery after shorter treatment (4 months). In addition, we isolated and identified several single compounds, justicidin A, 5-methoxyjusticidin A, chinensinaphthol, retrochinensinaphthol methyl ether, and suchilactone, from M. angustifolia and tested these compounds. Among them, justicidin A potently decreased the formation of Aβ in APPsw-transfected cells. These data suggest that the 60% EtOH extract of M. angustifolia has the potential to be developed as a treatment of AD. Furthermore, justicidin A may contribute, at least partially, to the Aβ alteration observed with the extract treatment.