MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cholecystokinin is up-regulated in obese mouse islets and expands beta-cell mass by increasing beta-cell survival.
Cholecystokinin is up-regulated in obese mouse islets and expands beta-cell mass by increasing beta-cell survival.
Endocrinology (2010-06-11)
Jeremy A Lavine, Philipp W Raess, Donald S Stapleton, Mary E Rabaglia, Joshua I Suhonen, Kathryn L Schueler, James E Koltes, John A Dawson, Brian S Yandell, Linda C Samuelson, Margery C Beinfeld, Dawn Belt Davis, Marc K Hellerstein, Mark P Keller, Alan D Attie
ABSTRACT
An absolute or functional deficit in beta-cell mass is a key factor in the pathogenesis of diabetes. We model obesity-driven beta-cell mass expansion by studying the diabetes-resistant C57BL/6-Leptin(ob/ob) mouse. We previously reported that cholecystokinin (Cck) was the most up-regulated gene in obese pancreatic islets. We now show that islet cholecystokinin (CCK) is up-regulated 500-fold by obesity and expressed in both alpha- and beta-cells. We bred a null Cck allele into the C57BL/6-Leptin(ob/ob) background and investigated beta-cell mass and metabolic parameters of Cck-deficient obese mice. Loss of CCK resulted in decreased islet size and reduced beta-cell mass through increased beta-cell death. CCK deficiency and decreased beta-cell mass exacerbated fasting hyperglycemia and reduced hyperinsulinemia. We further investigated whether CCK can directly affect beta-cell death in cell culture and isolated islets. CCK was able to directly reduce cytokine- and endoplasmic reticulum stress-induced cell death. In summary, CCK is up-regulated by islet cells during obesity and functions as a paracrine or autocrine factor to increase beta-cell survival and expand beta-cell mass to compensate for obesity-induced insulin resistance.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-PCNA (Ab-1) Mouse mAb (PC10), liquid, clone PC10, Calbiochem®