MilliporeSigma
  • Reactions of hypoiodous acid with model compounds and the formation of iodoform in absence/presence of permanganate.
Reactions of hypoiodous acid with model compounds and the formation of iodoform in absence/presence of permanganate.
Water research (2017-04-30)
Xiaodan Zhao, Jun Ma, Urs von Gunten
ABSTRACT
The kinetics for the reactions of hypoiodous acid (HOI) with various phenols (phenol, 4-nitrophenol, 4-hydroxybenzoic acid), 3-oxopentanedioic acid (3-OPA) and flavone were investigated in the pH range of 6.0-11.0. The apparent second order rate constants for the reactions of HOI with phenolic compounds, 3-OPA, flavone and citric acid at pH 8.0 are 10-10
Sigma-Aldrich
Flavone, ≥99.0%