MilliporeSigma
One-reactor plasma assisted fabrication of ZnO@TiO
Scientific reports (2017-08-31)
Alejandro Nicolas Filippin, Manuel Macias-Montero, Zineb Saghi, Jesús Idígoras, Pierre Burdet, Juan R Sanchez-Valencia, Angel Barranco, Paul A Migdley, Juan A Anta, Ana Borras
ABSTRACT
This paper addresses the fabrication of vertically aligned ZnO@TiO
Sigma-Aldrich
29H,31H-Phthalocyanine, β-form, 98%