MilliporeSigma
  • Highly selective enrichment of phosphopeptides by titanium (IV) attached monodisperse-porous poly(vinylphosphonic acid-co-ethylene dimethacrylate) microspheres.
Highly selective enrichment of phosphopeptides by titanium (IV) attached monodisperse-porous poly(vinylphosphonic acid-co-ethylene dimethacrylate) microspheres.
Journal of chromatography. A (2017-03-30)
Kouroush Salimi, Duygu Deniz Usta, Ömür Çelikbıçak, Aslı Pınar, Bekir Salih, Ali Tuncel
ABSTRACT
A seeded polymerization protocol was developed for the synthesis of monodisperse-porous poly(vinylphosphonic acid-co-ethylene dimethacrylate), [poly(VPA-co-EDMA)] microspheres with superior porous properties. The protocol allowed the direct synthesis of phosphonic acid functionalized porous microspheres with the mean size of ∼4μm and the specific surface area of 420m
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor