MilliporeSigma
  • Application of Ultrasound-Assisted Surfactant-Enhanced Emulsification Microextraction Based on Solidification of Floating Organic Droplets and High Performance Liquid Chromatography for Preconcentration and Determination of Alprazolam and Chlordiazepoxide in Human Serum Samples.
Application of Ultrasound-Assisted Surfactant-Enhanced Emulsification Microextraction Based on Solidification of Floating Organic Droplets and High Performance Liquid Chromatography for Preconcentration and Determination of Alprazolam and Chlordiazepoxide in Human Serum Samples.
Journal of chromatographic science (2017-03-24)
Nasser Goudarzi, Monavar Amirnavaee, Mansour Arab Chamjangali, Sahar Farsimadan
ABSTRACT
An improved microextraction method is proposed on the basis of ultrasound-assisted surfactant-enhanced emulsification and solidification of a floating organic droplet procedure combined with high performance liquid chromatography for the preconcentration and quantification of alprazolam (ALP) and chlordiazepoxide (CHL) present in a number of human serum samples. Several parameters affecting the extraction efficiency were investigated by the Plackett -Burman factorial design as the screening design. Then the response surface methodology based on the Box-Behnken design was used to optimize the effective parameters in the proposed procedure. The limits of detection for the proposed method were found to be 3.0 and 3.1 ng mL-1 for CHL and ALP, respectively. The calibration curves obtained for the method were linear in the ranges of 10.0-3,500.0 and 10.0-3,000.0 ng mL-1 for CHL and ALP, respectively, with a good determination coefficient. The recoveries of the drugs in the spiked human serum samples were above 93.0%. The developed method was successfully applied to the analysis of these studied drugs in human serum samples. The pre-treatment of the serum samples was performed using acetonitrile to remove the proteins. The proposed procedure was an accurate and reliable one for the determination and preconcentration of these drugs in blood samples.
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)