MilliporeSigma
  • Diffusion Kurtosis Imaging Study of Rectal Adenocarcinoma Associated with Histopathologic Prognostic Factors: Preliminary Findings.
Diffusion Kurtosis Imaging Study of Rectal Adenocarcinoma Associated with Histopathologic Prognostic Factors: Preliminary Findings.
Radiology (2016-12-09)
Lan Zhu, Zilai Pan, Qian Ma, Wenjie Yang, Hongyuan Shi, Caixia Fu, Xu Yan, Lianjun Du, Fuhua Yan, Huan Zhang
ABSTRACT
Purpose To determine the correlation between diffusion kurtosis imaging (DKI)-derived parameters and prognostic factors for rectal adenocarcinoma. Materials and Methods This study was approved by the local institute review board, and written informed consent was obtained from each patient. Data from 56 patients (median age, 59.5 years; age range, 31-86 years) with rectal adenocarcinoma between April 2014 and September 2015 were involved in this prospective study. DKI (b = 0, 700, 1400, and 2100 sec/mm
Sigma-Aldrich
Pyridinium dichromate, 98%