MilliporeSigma
  • Photo-activation of the delocalized lipophilic cation D112 potentiates cancer selective ROS production and apoptosis.
Photo-activation of the delocalized lipophilic cation D112 potentiates cancer selective ROS production and apoptosis.
Cell death & disease (2017-02-06)
Ning Yang, Michael Weinfeld, Hélène Lemieux, Ben Montpetit, Ing Swie Goping
ABSTRACT
Delocalized lipophilic cations (DLCs) selectively accumulate in cancer cell mitochondria and have long been explored for therapeutic applications. Although targeted effects to cancer cells are demonstrated in vitro, non-specific toxicities in vivo have hampered clinical development. Identifying the molecular mechanisms of action and enhancing selectivity are thus necessary next steps to improve these compounds and evaluate their suitability for further drug development. D112 is one such DLC with promising properties. We previously demonstrated that D112 selectively induced intrinsic apoptosis in transformed versus non-transformed cell lines. Here we show that D112 preferentially entered transformed cells where it interacted with, and damaged mitochondrial DNA, inhibited Complex I respiration and induced reactive oxygen species (ROS). ROS production was critical for Bax activation and subsequent apoptosis. Importantly, photo-activation of D112 potentiated selective ROS production and increased the window of toxicity towards cancer cells over non-transformed cells. Thus photodynamic therapy would be an exciting adjunct to D112 studies and may be generally applicable for other DLCs that are currently under therapeutic investigation.