MilliporeSigma
  • Controlling the Regioselectivity of Baeyer-Villiger Monooxygenases by Mutation of Active-Site Residues.
Controlling the Regioselectivity of Baeyer-Villiger Monooxygenases by Mutation of Active-Site Residues.
Chembiochem : a European journal of chemical biology (2017-05-16)
Kathleen Balke, Marcus Bäumgen, Uwe T Bornscheuer
ABSTRACT
Baeyer-Villiger monooxygenase (BVMO)-mediated regiodivergent conversions of asymmetric ketones can lead to the formation of "normal" or "abnormal" lactones. In a previous study, we were able to change the regioselectivity of a BVMO by mutation of the active-site residues to smaller amino acids, which thus created more space. In this study, we demonstrate that this method can also be used for other BVMO/substrate combinations. We investigated the regioselectivity of 2-oxo-Δ