MilliporeSigma
  • Modeling Drug-Induced Neuropathy Using Human iPSCs for Predictive Toxicology.
Modeling Drug-Induced Neuropathy Using Human iPSCs for Predictive Toxicology.
Clinical pharmacology and therapeutics (2016-11-20)
R Ohara, K Imamura, F Morii, N Egawa, K Tsukita, T Enami, R Shibukawa, T Mizuno, M Nakagawa, H Inoue
ABSTRACT
Drugs under development can cause unpredicted toxicity in humans due to differential drug responsiveness between humans and other disease models, resulting in clinical trial failures. Human induced pluripotent stem cells (iPSCs) are expected to represent a useful tool for toxicity testing. However, among many assays, appropriate cellular assays for predicting neurotoxicity in an iPSC-based model are still uncertain. Here we generated neurons from iPSCs of Charcot-Marie-Tooth disease (CMT) patients. Some CMT patients are sensitive to anticancer drugs and present with an adverse reaction of neuropathy. We analyzed cellular phenotypes and found that mitochondria in neurites of CMT neurons were morphologically shorter and showed slower mobility compared to control. A neurosphere assay showed that treatment with drugs known to cause neuropathy caused mitochondrial aggregations in neurites with adenosine triphosphate shortage in both CMT and control neurons, although more severely in CMT. These findings suggest that the genetically susceptible model could provide a useful tool to predict drug-induced neurotoxicity.