MilliporeSigma
  • Performance of a highly lipophilic Li+ lonophore in solvent polymeric membranes
Performance of a highly lipophilic Li+ lonophore in solvent polymeric membranes
Mikrochimica Acta (1990)
Bochenska, M., Simon, W.