MilliporeSigma
Multivariate Analysis of
Frontiers in neuroinformatics (2017-04-21)
Fermín Segovia, Juan M Górriz, Javier Ramírez, Francisco J Martínez-Murcia, Johannes Levin, Madeleine Schuberth, Matthias Brendel, Axel Rominger, Kai Bötzel, Gaëtan Garraux, Christophe Phillips
ABSTRACT
An early and differential diagnosis of parkinsonian syndromes still remains a challenge mainly due to the similarity of their symptoms during the onset of the disease. Recently,