MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Transforming activity and therapeutic targeting of C-terminal-binding protein 2 in Apc-mutated neoplasia.
Transforming activity and therapeutic targeting of C-terminal-binding protein 2 in Apc-mutated neoplasia.
Oncogene (2017-04-18)
E T Sumner, A T Chawla, A D Cororaton, J E Koblinski, R C Kovi, I M Love, B B Szomju, S Korwar, K C Ellis, S R Grossman
ABSTRACT
Overexpression of the transcriptional coregulators C-terminal binding proteins 1 and 2 (CtBP1 and 2) occurs in many human solid tumors and is associated with poor prognosis. CtBP modulates oncogenic gene expression programs and is an emerging drug target, but its oncogenic role is unclear. Consistent with this oncogenic potential, exogenous CtBP2 transformed primary mouse and human cells to anchorage independence similarly to mutant H-Ras. To investigate CtBP's contribution to in vivo tumorigenesis, Apc
Sigma-Aldrich
Suplatast tosylate, ≥98% (HPLC)