MilliporeSigma
  • Cardiac Outcomes After Ischemic Stroke or Transient Ischemic Attack: Effects of Pioglitazone in Patients With Insulin Resistance Without Diabetes Mellitus.
Cardiac Outcomes After Ischemic Stroke or Transient Ischemic Attack: Effects of Pioglitazone in Patients With Insulin Resistance Without Diabetes Mellitus.
Circulation (2017-03-02)
Lawrence H Young, Catherine M Viscoli, Jeptha P Curtis, Silvio E Inzucchi, Gregory G Schwartz, Anne M Lovejoy, Karen L Furie, Mark J Gorman, Robin Conwit, J Dawn Abbott, Daniel L Jacoby, Daniel M Kolansky, Steven E Pfau, Frederick S Ling, Walter N Kernan
ABSTRACT
Insulin resistance is highly prevalent among patients with atherosclerosis and is associated with an increased risk for myocardial infarction (MI) and stroke. The IRIS trial (Insulin Resistance Intervention after Stroke) demonstrated that pioglitazone decreased the composite risk for fatal or nonfatal stroke and MI in patients with insulin resistance without diabetes mellitus, after a recent ischemic stroke or transient ischemic attack. The type and severity of cardiac events in this population and the impact of pioglitazone on these events have not been described. We performed a secondary analysis of the effects of pioglitazone, in comparison with placebo, on acute coronary syndromes (MI and unstable angina) among IRIS participants. All potential acute coronary syndrome episodes were adjudicated in a blinded fashion by an independent clinical events committee. The study cohort was composed of 3876 IRIS participants, mean age 63 years, 65% male, 89% white race, and 12% with a history of coronary artery disease. Over a median follow-up of 4.8 years, there were 225 acute coronary syndrome events, including 141 MIs and 84 episodes of unstable angina. The MIs included 28 (19%) with ST-segment elevation. The majority of MIs were type 1 (94, 65%), followed by type 2 (45, 32%). Serum troponin was 10× to 100× upper limit of normal in 49 (35%) and >100× upper limit of normal in 39 (28%). Pioglitazone reduced the risk of acute coronary syndrome (hazard ratio, 0.71; 95% confidence interval, 0.54-0.94; Among patients with insulin resistance without diabetes mellitus, pioglitazone reduced the risk for acute coronary syndromes after a recent cerebrovascular event. Pioglitazone appeared to have its most prominent effect in preventing spontaneous type 1 MIs. URL: http://clinicaltrials.gov. Unique identifier: NCT00091949.
Sigma-Aldrich
Pioglitazone
Pricing and availability is not currently available.