MilliporeSigma
  • Galectin-1-driven upregulation of SDF-1 in pancreatic stellate cells promotes pancreatic cancer metastasis.
Galectin-1-driven upregulation of SDF-1 in pancreatic stellate cells promotes pancreatic cancer metastasis.
Cancer letters (2017-03-25)
Dong Qian, Zipeng Lu, Qingcheng Xu, Pengfei Wu, Lei Tian, Liangtao Zhao, Baobao Cai, Jie Yin, Yang Wu, Kevin F Staveley-O'Carroll, Kuirong Jiang, Yi Miao, Guangfu Li
ABSTRACT
Galectin-1, mainly expressed in activated pancreatic stellate cells (PSCs), is involved in many important cancer-related processes. However, very little is known how Galectin-1 modulates PSCs and subsequently impacts pancreatic cancer cells (PCCs). Our chemokine antibody array and in vitro studies demonstrates that Galectin-1 induces secretion of stromal cell-derived factor-1(SDF-1) in PSCs by activating NF-κB signaling. The secreted SDF-1 increases migration and invasion of PCCs. Knockdown of Galectin-1 and inhibitor-mediated blockade of SDF-1 as well as its ligand CXCR4 and NF-κB verifies the findings. In vivo experiment by knockdown of Galectin-1 in PSCs further demonstrates the conclusion. Collectively, the present studies demonstrate that Galectin-1-driven production of SDF-1 in PSCs through activation of NF-κB promotes metastasis in PDAC, offering a potential target in the treatment of pancreatic cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human LGALS1