- TLR2 activation induces antioxidant defence in human monocyte-macrophage cell line models.
TLR2 activation induces antioxidant defence in human monocyte-macrophage cell line models.
Oncotarget (2017-09-15)
Iwona Karwaciak, Michal Gorzkiewicz, Grzegorz Bartosz, Lukasz Pulaski
PMID28903338
ABSTRACT
When monocytes are recruited to inflammation/infection sites, extravasate and differentiate into macrophages, they encounter increasing levels of oxidative stress, both from exogenous and endogenous sources. In this study, we aimed to determine whether there are specific biochemical mechanisms responsible for an increase in oxidative stress resistance in differentiating macrophages. We performed experiments on