MilliporeSigma
ADC
Journal of magnetic resonance imaging : JMRI (2017-03-10)
Lifang Wu, Pengju Xu, Shengxiang Rao, Li Yang, Caizhong Chen, Hao Liu, Caixia Fu, Mengsu Zeng
ABSTRACT
To explore the threshold of intravoxel incoherent motion (IVIM) parameters, apparent diffusion coefficient [ADC Institutional Review Board approval and informed consent were obtained for this prospective study. There were 30 patients undergoing 1.5T magnetic resonance imaging (MRI) with IVIM-DWI of 12 b values (0, 10, 20, 30, 40, 50, 70, 100, 200, 300, 500, 800 s/mm Median PFS was 99 days, within which 16 patients progressed. The threshold of ADC The ADC 1 Technical Efficacy: Stage 2 J. MAGN. RESON. IMAGING 2017;46:820-830.