MilliporeSigma
IgH isotype-specific B cell receptor expression influences B cell fate.
Proceedings of the National Academy of Sciences of the United States of America (2017-09-20)
Pei Tong, Alessandra Granato, Teng Zuo, Neha Chaudhary, Adam Zuiani, Seung Seok Han, Rakesh Donthula, Akritee Shrestha, Debattama Sen, Jennifer M Magee, Michael P Gallagher, Cees E van der Poel, Michael C Carroll, Duane R Wesemann
ABSTRACT
Ig heavy chain (IgH) isotypes (e.g., IgM, IgG, and IgE) are generated as secreted/soluble antibodies (sIg) or as membrane-bound (mIg) B cell receptors (BCRs) through alternative RNA splicing. IgH isotype dictates soluble antibody function, but how mIg isotype influences B cell behavior is not well defined. We examined IgH isotype-specific BCR function by analyzing naturally switched B cells from wild-type mice, as well as by engineering polyclonal
Sigma-Aldrich
Mouse IgE ELISA Kit