MilliporeSigma
  • Biochemical efficacy, molecular docking and inhibitory effect of 2, 3-dimethylmaleic anhydride on insect acetylcholinesterase.
Biochemical efficacy, molecular docking and inhibitory effect of 2, 3-dimethylmaleic anhydride on insect acetylcholinesterase.
Scientific reports (2017-10-04)
Kabrambam D Singh, Rajendra K Labala, Thiyam B Devi, Ningthoujam I Singh, Heisnam D Chanu, Sonia Sougrakpam, Bunindro S Nameirakpam, Dinabandhu Sahoo, Yallappa Rajashekar
ABSTRACT
Evolution of resistance among insects to action of pesticides has led to the discovery of several insecticides (neonicotinoids and organophosphates) with new targets in insect nervous system. Present study evaluates the mode of inhibition of acetylchlonesterase (AChE), biochemical efficacy, and molecular docking of 2,3-dimethylmaleic anhydride, against Periplaneta americana and Sitophilus oryzae. The knockdown activity of 2,3-dimethylmaleic anhydride was associated with in vivo inhibition of AChE. At KD