MilliporeSigma
  • Sensitive detection of PD-L1 expression on circulating epithelial tumor cells (CETCs) could be a potential biomarker to select patients for treatment with PD-1/PD-L1 inhibitors in early and metastatic solid tumors.
Sensitive detection of PD-L1 expression on circulating epithelial tumor cells (CETCs) could be a potential biomarker to select patients for treatment with PD-1/PD-L1 inhibitors in early and metastatic solid tumors.
Oncotarget (2017-10-27)
Dorothea Sonja Schott, Monika Pizon, Ulrich Pachmann, Katharina Pachmann
ABSTRACT
The current cancer research strongly focuses on immune therapies, where the PD-1, with its ligands plays an important role. It is known that PD-L1 is frequently up-regulated in a number of different cancers and the relevance of this pathway has been extensively studied and therapeutic approaches targeting PD-1 and PD-L1 have been developed. We used a non-invasive, real-time biopsy for determining PD-L1 and PD-L2 expression in CETCs of solid cancer patients. CETCs were determined from blood of 128 patients suffering from breast (72), prostate (27), colorectal (18) and lung (11) cancer. The number of vital CETCs and the expression of PD-L1 and PD-L2 were investigated using the maintrac PD-L1 expressing CETCs were detected in 94.5% of breast, 100% of prostate, 95.4% of colorectal and 82% of lung cancer patients whereas only 75% of breast cancer patients had PD-L2 positive CETCs. In the PD-L1 and PD-L2 expressing patients the cell fraction of PD-L1 positive CETCs is significantly higher than the fraction of PD-L2 positive CETCs (54.6% vs. 28.7%; p<0.001). Breast cancer patients with metastatic disease had significantly more PD-L1 positive CETCs as compared to patients without metastasis (median 75% vs. 61.1%; p<0.05). PD-L1 seems to be a major factor in immune evasion and is highly expressed on CETCs regardless of the type of cancer. Monitoring the frequency of PD-L1 positive CETCs could reflect individual patient's response for an anti-PD-1/PD-L1 therapy and may be a promising target of anticancer treatment.
Sigma-Aldrich
L-(−)-Glucose, ≥99%